Germanium standard solution, 10000 mg/L Ge in 5% nitric acid with hydrofluoric acid (max. 1%) (de Ge), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific

14410.AD 14410.AE 14410.AP
14410.ADEA 119 EUR
14410.AD 14410.AE 14410.AP
Germanium standard solution, 10000 mg/L Ge in 5% nitric acid with hydrofluoric acid (max. 1%) (de Ge), standard for ICP, Specpure®
Standards Cationic Standards Germanium Standards

Molecular Formula: Ge in 5% HNO₃/tr. HF
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085310


Certifications: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR