Cadmium plasma standard solutions, for ICP, Specpure®

Supplier: Thermo Fisher Scientific

13813.AD 13813.AE 13813.AP 14424.AE 14424.AP
13813.ADEA 62 EUR
13813.AD 13813.AE 13813.AP 14424.AE 14424.AP
Cadmium plasma standard solutions, for ICP, Specpure®
Standards Cationic Standards Cadmium Standards

Molecular Formula: Cd in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010914


Certifications: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR