Terbium triacetate hydrate ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Fisher Scientific

Synonyms: Terbium(III) acetate hydrate

14600.09 14600.18
14600.09EA 166 EUR
14600.09 14600.18
Terbium triacetate hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Terbium triacetate hydrate

Purity: 99.9% CAS No.: 100587-92-6 Molecular Formula: C6H9O6Tb.xH2O Formula Weight: 336.08(anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic MDL No.: MFCD00149233

Formula: C₆H₉O₆Tb·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149233
CAS Number: 100587-92-6
EINECS: 247-065-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR