Dipraseodymium trisulphate octahydrate ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Praseodymium(III) sulphate octahydrate

14607.14 14607.22
14607.14EA 66.7 EUR
14607.14 14607.22
Dipraseodymium trisulphate octahydrate ≥99.9% (REO, rare earth oxide basis)
Dipraseodymium trisulphate octahydrate

Purity: 99.9% CAS No.: 13510-41-3 Molecular Formula: Pr2(SO4)3.8H2O Formula Weight: 714.12 (570.00anhy) Physical Form: Crystalline Appearance: Green Odour: Odourless Merck Reference: 14,7712 MDL No.: MFCD00149832

Formula: O₁₂Pr₂S₃·8H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149832
CAS Number: 13510-41-3
EINECS: 233-622-8
Merck Index: 14,7712

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR