Gold ≥99.95% (metals basis), foil, Thickness: 0.25 mm (0.01 in)

Supplier: Thermo Fisher Scientific

14722.FF 14722.FI 14722.GH
14722.FFEA 994 EUR
14722.FF 14722.FI 14722.GH
Gold ≥99.95% (metals basis), foil, Thickness: 0.25 mm (0.01 in)
Gold

Purity: 99.95% CAS No.: 7440-57-5 Physical Form: ≈3g/25x25mm Odour: Odourless MDL No.: MFCD00003436

Formula: Au
Boiling Pt: 2800 °C (1013 hPa)
Melting Pt: 1063 °C
Density: 19,3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR