Graphite ≥99.9995% (metals basis), rods, Length: 152mm (6in), Ø 6.15mm (0.242in)

Supplier: Thermo Scientific

14738.KD
14738.KDEA 518 EUR
14738.KD
Graphite ≥99.9995% (metals basis), rods, Length: 152mm (6in), Ø 6.15mm (0.242in)
Graphite
Formula: C
MW: 12,01 g/mol
Melting Pt: 3827 °C
Density: 2,09…2.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-955-3
Merck Index: 12,04560

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR