Pumps and colorimetric tubes for gas measurements
Supplier: GASTEC
600 840 TG-1 27 25 26 107 GV-100S GV-100LS GV-110S 860 28
151-0050EA 267 EUR
151-0050 151-0060 151-0068 151-0023 151-0021 151-0022 151-0000 151-0064 151-0063 118-0080 118-0473 GAST28
Pumps and colorimetric tubes for gas measurements
Gas Detection Systems
Identify and measure specific gases with the sampling pump and gas tubes.
  • Ergonomically designed sure-grip shape, lightweight and corrosion resistant
  • Built-in tube tip breaker makes tip breaking easy, safe, and convenient
  • Integral flow-finish indicator lets you know when sampling is complete
Use the specially designed manual pump to draw a precise 100 ml air sample through a gas analyser tube.
The resulting length-of-stain colour change can be read directly off the tube's calibration scale, yielding an accurate concentration.
Crystal clear gasMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable