Passive gas detectors, Dositubes
Supplier: GASTEC
5DH 32D 81D 9D 152D 14D 17D 174D 2D 1DL 133D 132D 1D 3D 4D 5D 12D 8D 91D 151D 122DL 3DL
151-0089EA 103 EUR
151-0089 151-0084 151-0090 151-0093 151-0078 151-0076 151-0080 151-0079 151-0083 151-0082 151-0075 151-0074 151-0081 151-0086 151-0094 151-0088 151-0073 151-0091 151-0062 151-0077 151-0072 151-0087
Passive gas detectors, Dositubes
Gas Tubes
The dositubes are used to measure time-weighted average (TWA) gas concentrations over longer periods.
  • Compact, lightweight, and versatile (can be used anywhere)
  • Easy direct measuring and reading
  • No sampling pump necessary
The Dositubes use the principle of the natural diffusion of gas.
A Dositube is a glass tube of a specific inner diameter that is filled with a diffuser and chemical reagent in parallel that enable gas to disperse at a constant rate resulting in a longer discolouration and better demarcation. The passive Dositube can then beMore Product Information