VWR® TISAB II Solution (Total Ionic Strength Adjustment Buffer) for Fluoride Determinations
Supplier: VWR Chemicals
160847GEA 323 EUR
160847G
VWR® TISAB II Solution (Total Ionic Strength Adjustment Buffer) for Fluoride Determinations
Buffers Ionic Strength Adjustment Buffers
Total ionic strength adjustment buffer solution for use with selective ion electrodes.