Barium chloride 10% in aqueous solution

Supplier: VWR Chemicals
Warning

164735PEA 135 EUR
164735P
Barium chloride 10% in aqueous solution
Barium chloride
Formula: BaCl₂
MW: 208.24 g/mol
Boiling Pt: >100 °C (1013 hPa)
Melting Pt: 963
Storage Temperature: Ambient
MDL Number: MFCD00003445
CAS Number: 10361-37-2
EINECS: 233-788-1
UN: 3287
ADR: 6.1 III
Merck Index: 12,00998

Order Now

Specification Test Results

Assay (W/V) 9.95 - 10.00 %
pH (25°C) 6.5 - 7.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR