Affinity chromatography columns, HiTrap™ protein G HP

Supplier: Cytiva
17-0404-03 17-0404-01 17-0405-01 17-0405-03 29-0485-81
17-0404-03EA 446 EUR
17-0404-03 17-0404-01 17-0405-01 17-0405-03 29-0485-81
Affinity chromatography columns, HiTrap™ protein G HP
Chromatography Columns
HiTrap™ Protein G HP affinity columns are pre-packed with 1 or 5 ml of Protein G Sepharose™ high performance for purification and isolation of monoclonal and polyclonal IgG from ascites, serum and cell culture supernatants.

  • Can be operated with a syringe, peristaltic pump or liquid chromatography system such as ÄKTAdesign™ or FPLC™ system
  • Polypropylene column material which is biocompatible and non interactive with biomolecules
  • Top and bottom frits are manufactured from porous polyethylene

Delivery information: Delivered with a stopper on the inletMore Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Max. pressure 5 bar
pH range 3 - 9
Storage 2 to 8 °C, 20% Ethanol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR