Hydrophobic interaction chromatography columns, HiTrap™ Capto™ Phenyl (High Sub)
Supplier: Cytiva
17-5451-09 17-5451-08
GEHE17-5451-09EA 545 EUR
GEHE17-5451-09 17-5451-08
Hydrophobic interaction chromatography columns, HiTrap™ Capto™ Phenyl (High Sub)
Chromatography Columns
Capto™ Phenyl (High sub) and Capto Butyl are hydrophobic interaction chromatography (HIC) resins used in the capture and intermediate stages of protein purification.
  • Aromatic ligand coupled with a flexible spacer for laboratory and process applications requiring a HIC resin with resin to high hydrophobicity
  • Highly rigid matrix enables high flow rates, long bed heights and faster throughput
  • Improves productivity and process economy in downstream operations
  • Narrow specification range ensures robust performance in sensitive processes
  • Excellent chemical
More Product Information