Water purification systems, RiOs-DI™ 3
Supplier: Merck
ZRDS0P3BE ZRDS0P3EU ZRDS0P3FR ZRDS0P3GE ZRDS0P3IT ZRDS0P3SP ZRDS0P3SZ ZRDSVP3IT ZRDSVP3SP ZRDSVP3SZ ZRDSVP3UK ZRDSVP3FR ZRDSVP3EU ZRDSVP3GE ZRDSVP3BE ZRDSVP3DK
171-1000EA 0 EUR
171-1000 171-1001 171-1002 171-1003 171-1004 171-1005 171-1006 171-1012 171-1013 171-1014 171-1015 171-1010 171-1009 171-1011 171-1008 MILLZRDSVP3DK
Water purification systems, RiOs-DI™ 3
Water Purification Systems Pure and Ultra Pure Water Systems
This easy-to-use RiOs-DI™ systems produce Type II pure water whose quality is superior to that of service DI or stills.
  • Combination of reverse osmosis (RO) technology and deionisation resins
  • Production of up to 15 L purified water per day; integrated 6 liter storage tank
  • Small footprint makes it easy to install the RiOs-DI® system wherever you want to in your lab
  • User-friendly display provides system status at a glance; the concise Quick Reference Guide is a handy guide for daily operation
  • Single SmartPak® RODI cartridge enables easy and rapid replacement
More Product Information