Anti-APP Rabbit Polyclonal Antibody

Supplier: Merck Millipore (Oncogene)
171609-50
171609-50EA 796 EUR
171609-50
Anti-APP Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR