Toluene ≥99.5% ACS, Riedel-de Haën™

Supplier: Honeywell Chemicals
Danger

Synonyms: Methylbenzene, Phenylmethane, FM approval

179418-5L
179418-5LEA 205 EUR
179418-5L
Toluene ≥99.5% ACS, Riedel-de Haën™
Toluene

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications


Beilstein Number: 635760

Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110.6 °C
Melting Pt: –95 °C
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3 II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Vapor Density 3.2 (vs air)
Assay (GC) Min. 99.50 %
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.03 %
S-compounds (as S) Max. 0.003 %
Reaction against H₂SO₄ complying
APHA Max. 10
IR spectrum complying
Vapor Pressure 26 mmHg ( 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR