Arsenic(III) oxide, Supelco®

Supplier: Merck
Danger

Synonyms: di-Arsenic trioxide

MQ100

17971-10G
17971-10GEA 88.6 EUR
17971-10G
Arsenic(III) oxide, Supelco®
Arsenic (III) oxide
Formula: As₂O₃
MW: 197.84 g/mol
Boiling Pt: 465
Melting Pt: 312
Density: 3.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003433
CAS Number: 1327-53-3
EINECS: 215-481-4
UN: 1561
ADR: 6.1 II
Merck Index: 12,00844
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f85e8ff5571c8ecb4261be61af6efc13632cd31&vid=786a9fdf133bdab95b63ddc73f9c65649909b8bd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR