Precast gels, CleanGel IEF
Supplier: Cytiva
18-1035-32
18-1035-32EA 561 EUR
18-1035-32
Precast gels, CleanGel IEF
Electrophoresis Gels Precast Gels
CleanGel IEF is a dried polyacrylamide gel optimised for analytical isoelectric focusing (IEF). The gel is excellent for separating enzymes with preserved activity.
  • Perform sensitive procedures using precast gels that are carefully washed and dried after casting to remove catalysts and unpolymerised material
  • Customise IEF separations by reswelling in a suitable mixture of carrier ampholytes and other additives such as detergents or denaturants
  • Handle mechanically stable, plastic-backed gels with confidence
The gel is cast on a polyester support filmMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable