Wash bottles, narrow neck
Supplier: KARTELL
179 180 182 183 185
179.EA 8.9 EUR
179. 180. 182. 183. 185.
Wash bottles, narrow neck
Bottles Wash Bottles
PE, translucent.
  • Flexible spout
  • Removable dispensing jet
  • Good chemical resistance
Narrow mouth wash bottles with screw cap.
Order Now
SPECIFICATIONS
Environmentally Preferable