Chemically-resistant liquid pumps LIQUIPORT®
Supplier: KNF
NF1.100FT.18S NF1.100KT.18RC NF1.100KT.18S NF1.100TT.18RC NF1.100TT.18S NF1.300FT.18RC NF1.300FT.18S NF1.300KT.18RC NF1.300KT.18S NF1.300TT.18RC NF1.300TT.18S NF100FT.18RC NF100FT.18S NF100KT.18RC NF100KT.18S NF100TT.18RC NF100TT.18S NF300FT.18RC NF300FT.18S NF300KT.18RC NF300KT.18S NF300TT.18RC NF300TT.18S NF1.100FT.18RC
224-0173EA 2860 EUR
224-0173 181-0128 181-0124 181-0129 181-0125 224-0174 224-0175 181-0136 181-0132 181-0137 181-0133 181-3923 181-3922 181-0126 181-0122 181-0127 181-0123 181-3925 181-3924 181-0134 181-0130 181-0135 181-0131 224-0172
Chemically-resistant liquid pumps LIQUIPORT®
Pumps Drum/Transfer Pumps
These diaphragm liquid pumps work quietly and with little vibration to pump neutral and aggressive liquids. Application: Transferring liquids.
  • Good chemical resistance
  • Self priming
  • Maintenance free, protected against dry running
  • Standby mode
  • Maximum pressure limiting
  • Flow rate from 0,2 up to 3 L/min/Pressure head 10 mWg, suction head 3 mWg
  • Pump heads available in PP, PVDF or PTFE, diaphragms available in PTFE, valves in FFKM
  • Pressure head also available for 60 mWg on request
  • Flow rate can either be set manually (Version S) or both
S version:
WithMore Product Information