Supelco® TraceCERT® Organic Reference Standard, Metazachlor Metabolite M479H008
Supplier: Merck
18164-10MG
18164-10MGEA 86.9 EUR
18164-10MG
Supelco® TraceCERT® Organic Reference Standard, Metazachlor Metabolite M479H008
Standards Organic Standards
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.