Vacuum butterfly valves, VS series
Supplier: VACUUBRAND
20665004 20665005 20665006 20665007 20665008 20665009
181-0155EA 433 EUR
181-0155 185-0001 VACU20665006 183-1131 183-1132 183-1133
Vacuum butterfly valves, VS series
Valves Vacuum Valves
The VS, VS C and VS B series valves are butterfly-type valves which offers very good conductance. The VS C series features a valve seal made of perfluorelastomer material (FFKM).
  • Compact, economic design
  • High conductance, low leakage rate
  • Gas-tight against atmosphere in both positions
  • Helium leak-tested
  • Easy to grip, lever-operated, with position indicator