Woulff bottles, DURAN®
Supplier: DWK Life Sciences
247104401 247105406 247107304
SCOT247104401EA 208 EUR
SCOT247104401 189-1110 SCOT247107304
Woulff bottles, DURAN®
Bottles Woulff Bottles
DURAN®, borosilicate glass 3.3, clear.
  • With bottom tubulature NS 19/26
  • Very good temperature resistance
  • Very good chemical resistance
Bottle with 3 grounded necks.
Certifications: DIN 12481.
Order Now
SPECIFICATIONS
Environmentally Preferable