Supelco® Organic Reference Standard, F.A.M.E. Mix Glc-30

Supplier: Merck

1893-1AMP
1893-1AMPEA 61.9 EUR
1893-1AMP
Supelco® Organic Reference Standard, F.A.M.E. Mix Glc-30
Standards Organic Standards

  • GHS signal: GHS07


Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.


Packaging: 100 mg ampule contains approximately equal amounts by weight of: Caprylic Acid Methyl Ester (C8:0) Capric Acid Methyl Ester (C10:0) Lauric Acid Methyl Ester (C12:0) Myristic Acid Methyl Ester (C14:0) Palmitic Acid Methyl Ester (C16:0)


Caution: P273 - P391 - P501

Order Now

SPECIFICATIONS

Synonym(s) Fatty Acid Methyl Ester Mix
Environmentally Preferable
M-Clarity quality level MQ100
Hazard class description H410
Shipping conditions DICE
Storage conditions –10...–25 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR