1-Butylpyridinium chloride ≥98%

Supplier: Thermo Fisher Scientific
Warning

19740.04 19740.09 19740.18
19740.04EA 60.8 EUR
19740.04 19740.09 19740.18
1-Butylpyridinium chloride ≥98%
1-Butylpyridinium chloride

Purity: 98% CAS No.: 1124-64-7 Molecular Formula: C9H14ClN Formula Weight: 171.67 Physical Form: Crystalline Sensitivity: Hygroscopic MDL No.: MFCD00060047

Formula: C₉H₁₄ClN
Melting Pt: 131…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00060047
CAS Number: 1124-64-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR