Sudan IV, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY

198102-25G
198102-25GEA 48.3 EUR
198102-25G
Sudan IV, Sigma-Aldrich®
Sudan IV

Sudan IV has been used to in en-face staining of aorta, stains the lipid rich plaques. It has also been used to stain lipids atherosclerotic lesions of mice.

Formula: C₂₄H₂₀N₄O
Melting Pt: 199 °C (dec., lit.)
Storage Temperature: Ambient
MDL Number: MFCD00003893
CAS Number: 85-83-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10ce9dd08992a21e0526a047ce197da922c95a7c&vid=a97ddaded4f6555d1480b2694694fca344d16c09&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR