Tin(II) chloride dihydrate 98-103% ACS

Supplier: VWR Chemicals
Danger

Synonyms: Stannous chloride dihydrate

1B1517-500GEA 432 EUR
1B1517-500G 1B1517-100G
Tin(II) chloride dihydrate 98-103% ACS
Tin (II) chloride dihydrate
Formula: SnCl₂·2H₂O
MW: 225.65 g/mol
Boiling Pt: 623
Melting Pt: 246
Density: 2.71 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149863
CAS Number: 10025-69-1
EINECS: 231-868-0
UN: 3260
ADR: 8 III
Merck Index: 13,08861

Order Now

Specification Test Results

Assay 98 - 103%
Calcium ≤0.005%
Iron ≤0.003%
Lead ≤0.01%
Potassium ≤0.005%
Sodium ≤0.01%
Solubility in hydrochloric acid Pass
Sulphate (P/F) Pass

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR