Flat flange lids with 4 necks, DURAN®
Supplier: DWK Life Sciences
243923406 243924608 243924702 243925107 243925707 243925801 243925904 243926009 243964603
273-0131EA 333 EUR
273-0131 201-0133 201-0134 201-0130 201-0135 201-0136 201-0137 201-0138 273-0132
Flat flange lids with 4 necks, DURAN®
Lids Reaction Vessel Lids
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For vacuum use. Flat flange (DN). Straight centre socket with NS 29/32 or NS 45/40. Maximum operating pressure at 250 °C: 1,0 bar.