Hydrogen peroxide 35% w/w water Stabilised analysis_for

Supplier: Thermo Fisher Scientific
Danger

202460010 202460025 202460250 202465000
202460010.EA 60 EUR
202460010. 202460025. 202460250. 202465000.
Hydrogen peroxide 35% w/w water Stabilised analysis_for
Hydrogen peroxide
Formula: H₂O₂
MW: 34.01 g/mol
Boiling Pt: ∼107 °C (1013 hPa)
Melting Pt: –33 °C
Density: 1.135 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00011333
CAS Number: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1 II
Merck Index: 14,04798

Order Now

Specification Test Results

Appearance Clear solution
Heavy metals ≤10 ppm
Color scale ≤10 APHA
Stabilizer about 0.03% stabilizers
Residue after evaporation ≤0.025 % (105°C)
Titration with KMnO4 34.5 to 37 %
Chloride (Cl) ≤2 ppm
Nitrates ≤0.012 %
Phosphates ≤0.012 %
Arsenic (As) ≤3 ppm
Chromium (Cr) ≤0.2 ppm
Iron (Fe) ≤0.5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR