Oxalic acid dihydrate 99, GPR RECTAPUR®

Supplier: VWR Chemicals
Warning

Synonyms: Ethanedioic acid dihydrate

20558.296EA 87.8 EUR
20558.296 20558.365 20558.410 20558.460
Oxalic acid dihydrate 99, GPR RECTAPUR®
Oxalic acid dihydrate
Formula: HO₂CCO₂H·2H₂O
MW: 126.07 g/mol
Boiling Pt: 149…160 °C (1013 hPa)
Melting Pt: 101
Density: 1.65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149102
CAS Number: 6153-56-6
EINECS: 205-634-3
UN: 3261
ADR: 8 III
REACH: 01-2119534576-33
Merck Index: 13,06980
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcc1447f5f23a589dc85e10dd090c86e33b6541b&vid=2f0e34543b2f625715b002905fc0dd45fcd75561&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99 %
Heavy metals (as Pb) Max. 10 ppm
Ignition residue (SO4) Max. 0.05 %
Fe (Iron) Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR