(±)-2-Butanol ≥98%, GPR RECTAPUR®

Supplier: VWR Chemicals
Warning

Synonyms: (±)-sec-Butanol, (±)-sec-Butyl alcohol, (±)-Ethyl methyl carbinol, (±)-2-Hydroxy butane, sec-Butanol, sec-Butyl alcohol, (±)-Methyl ethyl carbinol

20811.292EA 35.4 EUR
20811.292 20811.326
(±)-2-Butanol ≥98%, GPR RECTAPUR®
(±)-2-Butanol
Formula: H₃CCH₂CH(OH)CH₃
MW: 74.12 g/mol
Boiling Pt: 94
Melting Pt: –115 °C
Density: 0.8534 g/cm³ (20 °C)
Flash Pt: 23
Storage Temperature: Ambient
MDL Number: MFCD00004569
CAS Number: 78-92-2
EINECS: 201-158-5
UN: 1120
ADR: 3 III
Merck Index: 13,01540
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1dea3607167631fbfcf7751c20b71f345ad24c19&vid=010e04e8f508fd3d501873d20c2349817834b6e8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 98 %
Boiling point 98.5 - 100.5 °C
Density (20/4) 0.805 - 0.809
Evaporation residue Max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR