Ethanol, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

20824.296EA 36.9 EUR
20824.296 20824.321 20824.365 20824.467 20824.558 20824.930
Ethanol, GPR RECTAPUR®
Ethanol

Caution: WARNING : This alcohol is regulated. Please contact VWR. For duty paid orders in the UK please add DP to the product code.

Formula: C₂H₅OH
Boiling Pt: 78,3 °C (1013 hPa)
Melting Pt: –117 °C
Density: 0,7895 g/cm³ (20 °C)
Flash Pt: 12 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II
REACH: 01-2119457610-43

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.8 %
Assay (V/V) 96.0 - 96.3 %
Acidity Max. 0.0002 meq/g
Aldehydes (as CH3CHO) Max. 15 ppm
Evaporation residue Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR