Methanol, Wasser_0,005 99,8, AnalaR NORMAPUR® zS

Supplier: VWR Chemicals
Danger

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

20856.296EA 71.4 EUR
20856.296
Methanol, Wasser_0,005 99,8, AnalaR NORMAPUR® zS
Methanol
Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 (6.1) II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colourless liquid
Colour value Max. 10 APHA
Assay (calculated on anhydrous) Min. 99.8 %
Acidity (as HCOOH) Max. 0.0010 %
Alkalinity (as NH3) Max. 0.00010 %
Acetone + aldehydes (as CH3COCH3) Max. 0.0010 %
Formaldehyde Max. 0.0001 %
Residue on evaporation 0.0010 %
Reducing substances Max. 0.0002 %
Water (K.F.) Max. 0.0050 %
Substances discoloured by H2SO4 Max. 20 APHA
E/E Max. 0.100 %
Cl (Chloride) Max. 0.5 ppm
Al (Aluminium) Max. 0.5 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.5 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.01 ppm
Co (Cobalt) Max. 0.01 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.01 ppm
Fe (Iron) Max. 0.1 ppm
Mg (Magnesium) Max. 0.1 ppm
Mn (Manganese) Max. 0.01 ppm
Ni (Nickel) Max. 0.01 ppm
Pb (Lead) Max. 0.01 ppm
Sn (Tin) Max. 0.1 ppm
Zn (Zinc) Max. 0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR