F(ab’)2 Anti-IgG Goat Polyclonal Antibody (CF®750)

Supplier: Biotium
20920-50UL 20920-250UL
20920-50ULEA 103 EUR
20920-50UL 20920-250UL
F(ab’)2 Anti-IgG Goat Polyclonal Antibody (CF®750)
Antibodies
Goat anti-mouse IgG (H+L), F(ab’)2 fragment secondary antibody labeled with CF®750 dye.

Suitable for western, immunofluorescence, and immunohistology in FFPE tissues.

Type: Secondary
Antigen: Mouse IgG
Clonality: Polyclonal
Clone:
Conjugation: CF®750
Epitope:
Host: Goat
Isotype: IgG (H+L)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR