Vinylmagnesium bromide 0.7 M in THF, AcroSeal®

Supplier: Thermo Fisher Scientific
Danger

209391000 209398000
209391000.EA 68 EUR
209391000. 209398000.
Vinylmagnesium bromide 0.7 M in THF, AcroSeal®
Vinylmagnesium bromide

Appearance: Clear yellow to orange to brown Solution

Formula: C₂H₃BrMg
MW: 131.26 g/mol
Density: 0.98 g/cm³
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00000042
CAS Number: 1826-67-1
UN: 3399
ADR: 4.3 I

Order Now

Specification Test Results

Appearance Clear yellow to orange to brown Solution (May develop some turbidity or precipitate)
Titration Grignard 0.70 to 0.90 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR