Calcium chloride 0.1 mol/l (0.2 N) aqueous solution, Supelco®

Supplier: Merck

Synonyms: Chloro calcium, Calcium dichloride, CaCl2

MQ100

21059-250ML-F
21059-250ML-FEA 72.2 EUR
21059-250ML-F
Calcium chloride 0.1 mol/l (0.2 N) aqueous solution, Supelco®
Calcium chloride
Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: 1600
Melting Pt: 772
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
Merck Index: 14,01659

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR