Ammonium carbonate, AnalaR NORMAPUR® analytical reagent

Supplier: VWR Chemicals
Danger

21217.295EA 94.5 EUR
21217.295 21217.260
Ammonium carbonate, AnalaR NORMAPUR® analytical reagent
Ammonium carbonate

Mixture in approximatively equimolar ratio of ammonium carbonate NH4CO2NH2 and ammonium hydrogen carbonate NH4HCO3.

Formula: (NH₄)₂CO₃
Melting Pt: 58 °C
Density: 1,500 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010890
CAS Number: 10361-29-2
EINECS: 208-058-0
Merck Index: 13,00508

Order Now

Specification Test Results

Assay (NH3) 30.0 - 34.0 %
Heavy metals (as Pb) Max. 10 ppm
Ignition residue (SO4) Max. 50 ppm
Insolubility in water Max. 100 ppm
Cl (Chloride) Max. 5 ppm
NO3 (Nitrate) Max. 10 ppm
PO4 (Phosphate) Max. 10 ppm
SO4 (Sulphate) Max. 20 ppm
As (Arsenic) Max. 3 ppm
Fe (Iron) Max. 5 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR