Measuring beakers with handle, Nalgene®
Supplier: Thermo Fisher Scientific
1220-1250 1220-2500 1223-1000 1223-3000
213-7152EA 70.1 EUR
213-7152 213-7154 213-7162 213-7164
Measuring beakers with handle, Nalgene®
Beakers
HDPE, translucent or PMP (TPX), crystal clear.
  • Good chemical resistance (HDPE)/Very good chemical resistance (PMP)
  • Good thermal resistance (HDPE)/Heat resistant up to +150 °C (briefly to +175 °C) (PMP)
  • Designed for rough handling (HDPE)/Autoclavable (PMP)
Dual graduations in ml and oz. Square handle for easy carrying and pouring. Extra strong wall for safer handling of acids, bases and other solutions. Transparent to microwaves (PMP).