Ammonium thiocyanate ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent

Supplier: VWR Chemicals
Warning

21344.237EA 78.3 EUR
21344.237 21344.294 21344.460
Ammonium thiocyanate ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Ammonium thiocyanate
Formula: NH₄SCN
Melting Pt: 150 °C
Density: 1,305 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011428
CAS Number: 1762-95-4
EINECS: 217-175-6
Merck Index: 13,00564

Order Now

Specification Test Results

Assay Min. 98.5 %
pH (20°C; 5 %) 4.2 - 5.8
Heavy metals (as Pb) Max. 5 ppm
Ignition residue (SO4) Max. 0.025 %
Insolubility in water Max. 50 ppm
Substances reducing iodine Max. 0.4 %
Cl (Chloride) Max. 50 ppm
S (Sulphide) Max. 10 ppm
Cu (Copper) Max. 4 ppm
Fe (Iron) Max. 1 ppm
Pb (Lead) Max. 4 ppm
Conforms to Reag. Ph.Eur. Passes test
SO4 (Sulphate) Max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR