Erlenmeyer flasks, disposable, with screw cap, Nalgene®
Supplier: Thermo Fisher Scientific
4112-0250 4112-0500 4112-2000 4112-0125 4112-1000
214-0137EA 215 EUR
214-0137 214-0138 214-0139 214-1925 214-1926
Erlenmeyer flasks, disposable, with screw cap, Nalgene®
Flasks Erlenmeyer Flasks
PETG, transparent, with HDPE screw cap.
  • Leakproof HDPE screw cap closure can be opened partially for venting by twisting a quarter turn
  • Packaged individually for easy storage and handling
  • Reduced risk of cross-contamination
Sterile, disposable Erlenmeyer flasks reduce the risk of cross-contamination. Ideal for shakers and suspension cultures, and for creating or storing media. Raised graduations for approximate reading of the contents.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification NAR Certificate of Retirement