Erlenmeyer flasks with thread, DURAN®
Supplier: DWK Life Sciences
218033653 218035457 218034452 218032451
214-0190EA 46.1 EUR
214-0190 201-0376 214-0457 214-0456
Erlenmeyer flasks with thread, DURAN®
Flasks Erlenmeyer Flasks
DURAN®, borosilicate glass 3.3, clear, without or with red PBT screw cap.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Volume graduations and labelling area. A PTFE-coated seal incorporated in the cap guarantees that the flasks are absolutely leakproof.
With printed Retrace Code (batch identification).
Order Now
SPECIFICATIONS
Environmentally Preferable