Vydac 214MS C4, HPLC columns
Supplier: Avantor
214MS5.1505EA 1560 EUR
214MS5.1505 214MS5205 214MS5405 214MS5.07515 214MS5.315 214MS5215 214MS5415 214MS5.325 214MS52 214MS54 214MS1022 214FSK1210MS 214GK52MS/N 214GK54MS/N 214GD52MS/N 214GD54MS/N
Vydac 214MS C4, HPLC columns
Chromatography Columns
These Vydac MS columns are a later development from Grace for the reversed phase HPLC separation of biomolecules. A proprietary surface treatment and bonding process give Vydac MS columns unique selectivity. A variety of reversed phases makes this product line suitable for the analysis of small peptides to large intact, undenatured proteins.
  • Five reversed-phase chemistries
  • Excellent peak shape with little or no TFA
  • High protein recoveries make scale-up easy
Lower capacity than C18 or C8, suitable for hydrophobic proteins or when minimal organicMore Product Information