Safety wash bottles for solvents, Nalgene®
Supplier: Thermo Fisher Scientific
2421-0500
215-0115EA 48 EUR
215-0115
Safety wash bottles for solvents, Nalgene®
Bottles Wash Bottles
HDPE, with red, fluorinated PP spray cap.
  • Outstanding resistance to many organic solvents, heat resistant up to 90 °C
  • Non-metallic vent valve keeps contents in the bottle, no leakage
  • With red label: 'Solvent Wash Bottle'
Fluorinated bottles provide better diffusion block and reduce absorption and permeation of solvents. The fluorination increases the shelf-life of the bottles and reduces material loss through permeation.