Bottle Carriers
Supplier: HUPFER METALLWERKE
7500942 7500943 7500944 7500945 7500946 7500947 7500948 7500949 7500950 7500951 7500952 7500953
215-0150EA 183 EUR
215-0150 215-0151 215-0152 215-0153 215-0154 215-0155 215-0160 215-0161 215-0162 215-0163 215-0164 215-0165
Bottle Carriers
Carriers Bottle Carriers/Containers
Stainless steel or steel, coated.
  • Four non slip feet
  • Sturdy carrying handle
  • Good chemical and temperature resistance
These bottle carriers are suitable for transporting and storing bottles.
Colour: Silver/white
Order Now
SPECIFICATIONS
Environmentally Preferable