Bottles, narrow neck, ungrounded neck, DURAN®
Supplier: DWK Life Sciences
211611707 211612403 211613605 211614404 211616302 211617307 211618603 211619102 211615409
SCOT211611707EA 14.8 EUR
SCOT211611707 215-1174 215-1175 215-1176 215-1178 215-1179 215-1418 SCOT211619102 2116154.
Bottles, narrow neck, ungrounded neck, DURAN®
Bottles Glass Bottles
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes