Bottles, narrow neck, with screw cap, Winchester type,
Supplier: Azpack
215-1945 215-1946 215-1947 215-1948 215-1949 215-1950 215-1943 215-1944 215-1953 215-1956 215-1957 215-1959 215-1960 215-1963 215-1965
215-1945EA 82.8 EUR
215-1945 215-1946 215-1947 215-1948 215-1949 215-1950 215-1943 215-1944 215-1953 215-1956 215-1957 215-1959 215-1960 215-1963 215-1965
Bottles, narrow neck, with screw cap, Winchester type,
Bottles Glass Bottles
Clear or amber soda-lime glass with fitted black urea caps and polythene cone inserts.
  • Good chemical and physical properties
  • For use in applications that are not associated with high thermal loads
  • Stable and robust
Excellent for sample collection and storing liquids. Amber coloured to protect light-sensitive materials.