Bottles, wide neck, amber, with GLS 80® thread and screw cap, DURAN®
Supplier: DWK Life Sciences
1160146 1160151 1160150 1160149 1160148 1160147 218666959 218663653
215-2065EA 114 EUR
215-2065 215-2070 215-2069 215-2068 215-2067 215-2066 215-2852 215-2960
Bottles, wide neck, amber, with GLS 80® thread and screw cap, DURAN®
Bottles Glass Bottles
DURAN®, borosilicate glass 3,3, amber, with blue PP screw cap.
  • Very good chemical resistance, high temperature resistance
  • Extra wide opening allows easy cleaning
  • Minimal thermal expansion giving relatively high resistance to temperature changes
  • Glass Type I according to USP, EP and JP
  • Amber glass meets USP 671 and Eur. Ph. 3.2.1. for protection against UV light
Amber to protect light-sensitive materials. UV protection up to approx. 500 nm light wavelength. Only the outer surface of the bottles is tinted ensuring that the DURAN® properties on theMore Product Information