Bottles, narrow neck, round, LDPE, with screw caps
Supplier: TECHNICAL TREATMENTS
016
215-2713EA 24.5 EUR
215-2713
Bottles, narrow neck, round, LDPE, with screw caps
Bottles Plastic Bottles
LDPE, natural colour.
  • Chemical resistant, black screw cap, leakproof
  • Good chemical resistance, good thermal resistance
  • With specially reinforced shoulder