Safety wash bottles, wide neck, multilingual, Azlon®
Supplier: SciLabware
WGW541PML WGW536PML WGW530PML WGW531PML WGW532PML WGW533PML WGW534PML WGW535PML WGW537PML WGW538PML WGW539PML WGW540PML WGW600PML WGW601PML WGW602PML WGW603PML WGW020PML
215-0417EA 39.8 EUR
215-0417 215-2796 215-2794 215-2885 215-2886 215-2887 215-2888 215-2795 215-2797 215-2889 215-2890 215-2891 215-2892 215-2798 215-2893 215-2894 215-3219
Safety wash bottles, wide neck, multilingual, Azlon®
Bottles Wash Bottles
LDPE, translucent, with PP spray cap.
  • Indelibly printed in four languages (English, French, German and Spanish)
  • Choose from three water or five solvent labels all including the new Globally Harmonised Symbols, NFPA diamond, risk phrases and safety information
  • Closure feature precision jet tip aperture for fine control
Wide mouth for easy, safe filling. Fitted with colour coded one-piece PP closure that maintain their shape in use.
Certifications: Azlon® wash bottles meet the material requirements of the U.S. Food and Drug Administration asMore Product Information