Safety wash bottles, wide neck, venting, multilingual, Azlon®
Supplier: SciLabware
WGW531VTML WGW532VTML WGW533VTML WGW534VTML WGW535VTML WGW537VTML WGW538VTML WGW539VTML WGW540VTML WGW541VTML WGW538VTMLR WGW539VTMLR WGW541VTMLR WGW537VTMLME WGW538VTMLME WGW539VTMLME WGW541VTMLME
215-3131EA 39.7 EUR
215-3131 215-3280 215-3281 215-3282 215-3132 215-3133 215-3283 215-3134 215-3284 215-3285 215-3870 215-3869 215-3868 215-3871 215-3874 215-3873 215-3872
Safety wash bottles, wide neck, venting, multilingual, Azlon®
Bottles Wash Bottles
LDPE, translucent with PP spray cap.
  • The patented DripLok® vapour venting valve prevents solvent drips
  • Indelibly printed in four languages (English, French, German and Spanish)
  • Closure feature precision jet tip aperture for fine control
Wide neck for easy, safe filling. Fitted with colour coded one-piece PP closure that maintains its shape in use.
Certifications: Azlon® wash bottles meet the material requirements of the U.S. Food and Drug Administration as specified in 21 CFR 177.1520.
Order Now
SPECIFICATIONS
Environmentally Preferable