DURAN® protect+ Laboratory Bottles, Plastic Safety Coated, DIN 168-1 Thread, Graduated, Amber
Supplier: DWK Life Sciences
218062487 218061482 218061782 218063689 218064488 218065484 218066386 218067382
215-4848EA 49.3 EUR
215-4848 215-4846 215-4847 215-4849 215-4850 215-4851 215-4852 215-4853
DURAN® protect+ Laboratory Bottles, Plastic Safety Coated, DIN 168-1 Thread, Graduated, Amber
Bottles Glass Bottles
Multi-functional ambered glass laboratory bottles with UV protection and a protective safety coating.
  • Manufactured from ambered borosilicate glass 3.3 and polyamide
  • Standardised GL-thread compatible with matching cap system for particularly tight sealing and simple, clean pouring out
  • The protect+ safety coating provides scratch, leak*, and splinter* protection (*only applies to bottles 2000 ml and less)
  • UV protection USP <660> (Spectral Transmission) and EP (3.2.1) compliant; less than 10% spectral transmission between 290 nm and 450 nm
  • External only
More Product Information